Assay Details
Target Gene Details
Entrez Gene ID: | 23246 |
Gene Name: | block of proliferation 1 |
Gene Aliases: |
- |
Location: |
Chr.8:144262046-144291438 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BOP1 | NM_015201.4 | NP_056016.1 | ||
AB060694.1 | BAB70666.1 | |||
AK024840.1 | ||||
AK290677.1 | ||||
AK293719.1 | ||||
AK315612.1 | ||||
BC001086.2 | AAH01086.2 | |||
BC005160.2 | AAH05160.2 | |||
BC007274.1 | AAH07274.1 | |||
BC013787.1 | AAH13787.1 | |||
BC013980.2 | AAH13980.1 | |||
BC017674.2 | AAH17674.1 | |||
BQ711345.1 | ||||
D50914.1 | BAA09473.1 | |||
DA510177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv12434n54 | Chr.8:144270080 - 144516801 on Build GRCh38 | Loss | CYHR1 ADCK5 FOXH1 FBXL6 HSF1 RECQL4 LOC101928902 MIR1234 PPP1R16A SCRT1 VPS28 MIR939 GPT CPSF1 LOC101928953 DGAT1 TMEM249 SLC39A4 MIR6849 TONSL-AS1 BOP1 MIR6893 MFSD3 MIR6848 TONSL SLC52A2 KIFC2 |
nsv470249 | Chr.8:143889098 - 144519234 on Build GRCh38 | Loss | CYHR1 EXOSC4 PARP10 ADCK5 MIR7112 FOXH1 MIR661 FBXL6 HGH1 LRRC14 PLEC HSF1 CYC1 RECQL4 LOC101928902 MIR1234 SHARPIN MIR6847 WDR97 PPP1R16A SCRT1 VPS28 GPAA1 MIR939 GPT CPSF1 LOC101928953 DGAT1 OPLAH TMEM249 SLC39A4 MIR6849 TONSL-AS1 BOP1 MAF1 MIR6893 MROH1 SPATC1 MFSD3 MIR6848 MIR6846 SCX TONSL SMPD5 SLC52A2 GRINA KIFC2 |
nsv515809 | Chr.8:143904439 - 144519234 on Build GRCh38 | Gain+Loss | CYHR1 EXOSC4 PARP10 ADCK5 MIR7112 FOXH1 MIR661 FBXL6 HGH1 LRRC14 PLEC HSF1 CYC1 RECQL4 LOC101928902 MIR1234 SHARPIN MIR6847 WDR97 PPP1R16A SCRT1 VPS28 GPAA1 MIR939 GPT CPSF1 LOC101928953 DGAT1 OPLAH TMEM249 SLC39A4 MIR6849 TONSL-AS1 BOP1 MAF1 MIR6893 MROH1 SPATC1 MFSD3 MIR6848 MIR6846 SCX TONSL SMPD5 SLC52A2 GRINA KIFC2 |
nsv1025071 | Chr.8:144044224 - 144300847 on Build GRCh38 | Gain | EXOSC4 MIR7112 OPLAH HGH1 BOP1 HSF1 CYC1 MAF1 MROH1 SPATC1 SHARPIN MIR6846 MIR6847 SCX WDR97 SMPD5 GPAA1 |
nsv428208 | Chr.8:144041826 - 144722291 on Build GRCh38 | Gain | LRRC24 CYHR1 EXOSC4 ADCK5 MIR7112 FOXH1 ZNF251 FBXL6 HGH1 LRRC14 HSF1 CYC1 RECQL4 LOC101928902 MIR1234 SHARPIN MIR6847 WDR97 PPP1R16A SCRT1 VPS28 GPAA1 MIR939 GPT CPSF1 LOC101928953 C8orf82 DGAT1 OPLAH TMEM249 SLC39A4 MIR6849 TONSL-AS1 BOP1 MAF1 MIR6893 MROH1 SPATC1 MFSD3 ARHGAP39 MIR6848 MIR6846 SCX TONSL SMPD5 SLC52A2 KIFC2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |