Assay Details
Target Gene Details
Entrez Gene ID: | 114569 |
Gene Name: | mal, T-cell differentiation protein 2 (gene/pseudogene) |
Gene Aliases: |
- |
Location: |
Chr.8:119208371-119245673 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAL2 | NM_052886.2 | NP_443118.1 | ||
XM_011516807.2 | XP_011515109.1 | |||
AK172820.1 | BAD18789.1 | |||
AK312030.1 | ||||
AY007723.1 | AAG15576.1 | |||
BC012367.1 | AAH12367.1 | |||
CV574204.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759639 | Chr.8:118931070 - 119263510 on Build GRCh38 | Gain | COLEC10 LOC101927513 TNFRSF11B MAL2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |