Assay Details
Target Gene Details
Entrez Gene ID: | 7991 |
Gene Name: | tumor suppressor candidate 3 |
Gene Aliases: |
D8S1992, M33, MRT22, MRT7, N33, OST3A |
Location: |
Chr.8:15540087-15766649 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TUSC3 | NM_006765.3 | NP_006756.2 | ||
NM_178234.2 | NP_839952.1 | |||
BC010370.1 | AAH10370.1 | |||
BT020002.1 | AAV38805.1 | |||
DA796279.1 | ||||
U42349.1 | AAB18374.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1161734 | Chr.8:15533647 - 15548772 on Build GRCh38 | Deletion | TUSC3 |
nsv437055 | Chr.8:15398926 - 15806828 on Build GRCh38 | Loss | TUSC3 |
nsv6089 | Chr.8:15538984 - 15565828 on Build GRCh38 | Deletion | TUSC3 |
esv2759598 | Chr.8:14768691 - 15907197 on Build GRCh38 | Loss | MIR383 TUSC3 SGCZ |
dgv248e55 | Chr.8:14773418 - 15806518 on Build GRCh38 | Loss | MIR383 TUSC3 SGCZ |
dgv12015n54 | Chr.8:15544498 - 15551464 on Build GRCh38 | Loss | TUSC3 |
dgv155e203 | Chr.8:15542279 - 15555938 on Build GRCh38 | Loss | TUSC3 |
esv3616406 | Chr.8:15544155 - 15557282 on Build GRCh38 | Loss | TUSC3 |
esv3543118 | Chr.8:15544264 - 15561491 on Build GRCh38 | Deletion | TUSC3 |
nsv1029399 | Chr.8:14978756 - 16296799 on Build GRCh38 | Gain | TUSC3 SGCZ MSR1 |
nsv8298 | Chr.8:14772387 - 15806223 on Build GRCh38 | Gain+Loss | MIR383 TUSC3 SGCZ |
nsv465554 | Chr.8:15537635 - 15554099 on Build GRCh38 | Loss | TUSC3 |
nsv824551 | Chr.8:15544358 - 15556098 on Build GRCh38 | Loss | TUSC3 |
esv28500 | Chr.8:15544336 - 15555629 on Build GRCh38 | Loss | TUSC3 |
esv3616403 | Chr.8:15528780 - 15600263 on Build GRCh38 | Loss | TUSC3 |
nsv517314 | Chr.8:15531057 - 15554099 on Build GRCh38 | Loss | TUSC3 |
nsv1161736 | Chr.8:15543669 - 15555938 on Build GRCh38 | Deletion | TUSC3 |
esv3616407 | Chr.8:15544186 - 15580033 on Build GRCh38 | Loss | TUSC3 |
nsv610466 | Chr.8:14469128 - 15809390 on Build GRCh38 | Loss | MIR383 TUSC3 SGCZ |
nsv818605 | Chr.8:14773392 - 15802730 on Build GRCh38 | Loss | MIR383 TUSC3 SGCZ |
nsv610524 | Chr.8:15419198 - 15568674 on Build GRCh38 | Loss | TUSC3 |
nsv1161735 | Chr.8:15543415 - 15565761 on Build GRCh38 | Deletion | TUSC3 |
dgv2048e212 | Chr.8:15543403 - 15555926 on Build GRCh38 | Loss | TUSC3 |
dgv12014n54 | Chr.8:15543065 - 15555467 on Build GRCh38 | Loss | TUSC3 |
nsv1020054 | Chr.8:15444894 - 15812092 on Build GRCh38 | Gain | TUSC3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |