Assay Details
Target Gene Details
Entrez Gene ID: | 729330 |
Gene Name: | otoconin 90 |
Gene Aliases: |
PLA2L |
Location: |
Chr.8:132024220-132059380 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OC90 | NM_001080399.2 | NP_001073868.2 | ||
AK297911.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1018172 | Chr.8:131797977 - 132121114 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv612325 | Chr.8:131918447 - 132338873 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
esv3618853 | Chr.8:131375690 - 132136917 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)