Assay Details
Target Gene Details
Entrez Gene ID: | 23246 |
Gene Name: | block of proliferation 1 |
Gene Aliases: |
- |
Location: |
Chr.8:144262046-144291438 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BOP1 | NM_015201.4 | NP_056016.1 | ||
AB060694.1 | BAB70666.1 | |||
AK024840.1 | ||||
AK290677.1 | ||||
AK293719.1 | ||||
AK315612.1 | ||||
BC001086.2 | AAH01086.2 | |||
BC005160.2 | AAH05160.2 | |||
BC007274.1 | AAH07274.1 | |||
BC013787.1 | AAH13787.1 | |||
BC013980.2 | AAH13980.1 | |||
BC017674.2 | AAH17674.1 | |||
BQ711345.1 | ||||
D50914.1 | BAA09473.1 | |||
DA510177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv12434n54 | Chr.8:144270080 - 144516801 on Build GRCh38 | Loss | HSF1 MIR6849 KIFC2 SCRT1 CPSF1 TONSL MIR6893 ADCK5 MFSD3 LOC101928953 MIR1234 DGAT1 SLC39A4 MIR939 GPT RECQL4 LOC101928902 PPP1R16A FOXH1 CYHR1 FBXL6 SLC52A2 BOP1 TMEM249 MIR6848 VPS28 TONSL-AS1 |
nsv470249 | Chr.8:143889098 - 144519234 on Build GRCh38 | Loss | HSF1 PLEC SMPD5 MIR6849 KIFC2 MAF1 CYC1 SCRT1 PARP10 CPSF1 TONSL MIR6893 MROH1 OPLAH SCX MIR661 ADCK5 SPATC1 MIR7112 MFSD3 LRRC14 GPAA1 WDR97 LOC101928953 MIR1234 DGAT1 SLC39A4 MIR939 GPT RECQL4 LOC101928902 PPP1R16A SHARPIN FOXH1 HGH1 GRINA MIR6847 CYHR1 FBXL6 MIR6846 SLC52A2 EXOSC4 BOP1 TMEM249 MIR6848 VPS28 TONSL-AS1 |
nsv515809 | Chr.8:143904439 - 144519234 on Build GRCh38 | Gain+Loss | HSF1 PLEC SMPD5 MIR6849 KIFC2 MAF1 CYC1 SCRT1 PARP10 CPSF1 TONSL MIR6893 MROH1 OPLAH SCX MIR661 ADCK5 SPATC1 MIR7112 MFSD3 LRRC14 GPAA1 WDR97 LOC101928953 MIR1234 DGAT1 SLC39A4 MIR939 GPT RECQL4 LOC101928902 PPP1R16A SHARPIN FOXH1 HGH1 GRINA MIR6847 CYHR1 FBXL6 MIR6846 SLC52A2 EXOSC4 BOP1 TMEM249 MIR6848 VPS28 TONSL-AS1 |
nsv1025071 | Chr.8:144044224 - 144300847 on Build GRCh38 | Gain | HSF1 SMPD5 MAF1 CYC1 SHARPIN MROH1 HGH1 OPLAH MIR6847 SCX MIR6846 SPATC1 EXOSC4 BOP1 MIR7112 GPAA1 WDR97 |
nsv428208 | Chr.8:144041826 - 144722291 on Build GRCh38 | Gain | HSF1 SMPD5 MIR6849 KIFC2 MAF1 LRRC24 CYC1 SCRT1 CPSF1 TONSL MIR6893 MROH1 OPLAH SCX ADCK5 C8orf82 SPATC1 MIR7112 MFSD3 LRRC14 GPAA1 WDR97 LOC101928953 ZNF251 MIR1234 DGAT1 SLC39A4 MIR939 GPT RECQL4 LOC101928902 PPP1R16A SHARPIN FOXH1 HGH1 MIR6847 CYHR1 FBXL6 MIR6846 SLC52A2 EXOSC4 BOP1 TMEM249 MIR6848 VPS28 TONSL-AS1 ARHGAP39 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |