Assay Details
Target Gene Details
Entrez Gene ID: | 51606 |
Gene Name: | ATPase H+ transporting V1 subunit H |
Gene Aliases: |
CGI-11, MSTP042, NBP1, SFD, SFDalpha, SFDbeta, VMA13 |
Location: |
Chr.8:53715543-53843311 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP6V1H | NM_015941.3 | NP_057025.2 | ||
NM_213619.2 | NP_998784.1 | |||
NM_213620.2 | NP_998785.1 | |||
XM_006716455.2 | XP_006716518.1 | |||
XM_011517542.1 | XP_011515844.1 | |||
AF112204.1 | AAF17192.1 | |||
AF113222.1 | AAG39293.1 | |||
AF132945.1 | AAD27720.1 | |||
AF298777.1 | AAG22809.1 | |||
AI741756.1 | ||||
AK022345.1 | ||||
AK094839.1 | ||||
AK098305.1 | ||||
AK303229.1 | ||||
AK308204.1 | ||||
BC025275.1 | AAH25275.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759616 | Chr.8:53562029 - 53824913 on Build GRCh38 | Loss | ATP6V1H |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |