Assay Details
Target Gene Details
Entrez Gene ID: | 7532 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma |
Gene Aliases: |
14-3-3GAMMA, PPP1R170 |
Location: |
Chr.7:76326791-76359025 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YWHAG | NM_012479.3 | NP_036611.2 | ||
AB024334.1 | BAA85184.1 | |||
AF142498.1 | AAD48408.1 | |||
AK024230.1 | ||||
AK293505.1 | ||||
AK295029.1 | ||||
BC020963.2 | AAH20963.1 | |||
CR541904.1 | CAG46702.1 | |||
CR541925.1 | CAG46723.1 | |||
DC343104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422494 | Chr.7:76286900 - 76984449 on Build GRCh38 | Duplication | LOC100133091 SSC4D LOC105375355 SPDYE16 ZP3 HSPB1 FDPSP2 POMZP3 SRRM3 DTX2P1-UPK3BP1-PMS2P11 DTX2 YWHAG |
nsv428174 | Chr.7:76305019 - 77220062 on Build GRCh38 | Gain | LOC100133091 LOC105375355 ZP3 PMS2P9 DTX2P1-UPK3BP1-PMS2P11 DTX2 YWHAG SPDYE17 SSC4D SPDYE16 CCDC146 SPDYE18 FDPSP2 POMZP3 FGL2 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | NSUN5P1 LOC105375355 STYXL1 NCF1C STAG3L1 SRRM3 LOC107986711 CCL26 SSC4D GTF2IP1 LOC441259 FDPSP2 POMZP3 TMEM120A GTF2IRD2B SNORA14A POM121C LOC100133091 SPDYE5 CCL24 POR GTF2IP7 ZP3 MIR4651 MDH2 TRIM73 DTX2 RHBDD2 YWHAG PMS2P10 PMS2P3 HIP1 SPDYE16 PMS2P2 HSPB1 LOC541473 |
nsv1028665 | Chr.7:76336044 - 76445651 on Build GRCh38 | Gain | SSC4D ZP3 YWHAG |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | LOC105375355 STYXL1 SRRM3 LOC107986711 CCL26 SSC4D FDPSP2 POMZP3 TMEM120A SNORA14A POM121C LOC100133091 SPDYE5 CCL24 POR GTF2IP7 ZP3 MIR4651 MDH2 DTX2 RHBDD2 YWHAG PMS2P3 HIP1 SPDYE16 HSPB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |