Assay Details
Target Gene Details
Entrez Gene ID: | 7532 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma |
Gene Aliases: |
14-3-3GAMMA, PPP1R170 |
Location: |
Chr.7:76326791-76359025 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YWHAG | NM_012479.3 | NP_036611.2 | ||
AB024334.1 | BAA85184.1 | |||
AF142498.1 | AAD48408.1 | |||
AK024230.1 | ||||
AK293505.1 | ||||
AK295029.1 | ||||
BC020963.2 | AAH20963.1 | |||
CR541904.1 | CAG46702.1 | |||
CR541925.1 | CAG46723.1 | |||
DC343104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422494 | Chr.7:76286900 - 76984449 on Build GRCh38 | Duplication | POMZP3 SRRM3 LOC100133091 LOC105375355 DTX2 HSPB1 DTX2P1-UPK3BP1-PMS2P11 YWHAG SPDYE16 FDPSP2 ZP3 SSC4D |
nsv428174 | Chr.7:76305019 - 77220062 on Build GRCh38 | Gain | POMZP3 FGL2 CCDC146 LOC105375355 DTX2 DTX2P1-UPK3BP1-PMS2P11 YWHAG SPDYE16 FDPSP2 ZP3 LOC100133091 SPDYE18 SPDYE17 PMS2P9 SSC4D |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | POMZP3 RHBDD2 LOC105375355 GTF2IP7 DTX2 POM121C GTF2IRD2B PMS2P2 FDPSP2 STAG3L1 ZP3 SPDYE5 MDH2 SNORA14A CCL26 PMS2P3 SSC4D LOC441259 SRRM3 PMS2P10 NCF1C TRIM73 YWHAG SPDYE16 NSUN5P1 LOC541473 STYXL1 POR GTF2IP1 LOC100133091 LOC107986711 HSPB1 HIP1 MIR4651 CCL24 TMEM120A |
nsv1028665 | Chr.7:76336044 - 76445651 on Build GRCh38 | Gain | YWHAG ZP3 SSC4D |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | POMZP3 RHBDD2 LOC105375355 GTF2IP7 DTX2 POM121C FDPSP2 ZP3 SPDYE5 MDH2 SNORA14A CCL26 PMS2P3 SSC4D SRRM3 YWHAG SPDYE16 STYXL1 POR LOC100133091 LOC107986711 HSPB1 HIP1 MIR4651 CCL24 TMEM120A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |