Assay Details
Target Gene Details
Entrez Gene ID: | 25798 |
Gene Name: | brain protein I3 |
Gene Aliases: |
I3 |
Location: |
Chr.7:98281667-98322398 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BRI3 | NM_001159491.1 | NP_001152963.1 | ||
NM_015379.4 | NP_056194.1 | |||
XM_017011931.1 | XP_016867420.1 | |||
XM_017011932.1 | XP_016867421.1 | |||
XM_017011933.1 | XP_016867422.1 | |||
XM_017011934.1 | XP_016867423.1 | |||
XM_017011935.1 | XP_016867424.1 | |||
XM_017011936.1 | XP_016867425.1 | |||
AB055977.1 | BAB32785.1 | |||
AF041430.2 | AAF18565.2 | |||
AF106966.1 | AAD05167.1 | |||
BC018737.1 | AAH18737.1 | |||
BC062370.1 | AAH62370.1 | |||
BF108440.1 | ||||
BF432881.1 | ||||
BQ018487.1 | ||||
CB054862.1 | ||||
CB528682.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831067 | Chr.7:98230857 - 98436216 on Build GRCh38 | Loss | BAIAP2L1 BRI3 TECPR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |