Assay Details
Target Gene Details
Entrez Gene ID: | 7532 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma |
Gene Aliases: |
14-3-3GAMMA, PPP1R170 |
Location: |
Chr.7:76326791-76359025 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YWHAG | NM_012479.3 | NP_036611.2 | ||
AB024334.1 | BAA85184.1 | |||
AF142498.1 | AAD48408.1 | |||
AK024230.1 | ||||
AK293505.1 | ||||
AK295029.1 | ||||
BC020963.2 | AAH20963.1 | |||
CR541904.1 | CAG46702.1 | |||
CR541925.1 | CAG46723.1 | |||
DC343104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422494 | Chr.7:76286900 - 76984449 on Build GRCh38 | Duplication | DTX2P1-UPK3BP1-PMS2P11 YWHAG POMZP3 SPDYE16 LOC100133091 SRRM3 LOC105375355 SSC4D DTX2 HSPB1 FDPSP2 ZP3 |
nsv428174 | Chr.7:76305019 - 77220062 on Build GRCh38 | Gain | YWHAG POMZP3 FDPSP2 PMS2P9 DTX2P1-UPK3BP1-PMS2P11 SPDYE16 SPDYE17 LOC100133091 FGL2 CCDC146 LOC105375355 SSC4D DTX2 SPDYE18 ZP3 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | GTF2IRD2B POMZP3 HIP1 MDH2 MIR4651 CCL26 POR STYXL1 SPDYE16 NCF1C GTF2IP1 LOC541473 LOC105375355 SSC4D DTX2 SPDYE5 PMS2P3 YWHAG TRIM73 CCL24 RHBDD2 SNORA14A HSPB1 FDPSP2 TMEM120A PMS2P2 NSUN5P1 STAG3L1 LOC107986711 PMS2P10 LOC100133091 LOC441259 SRRM3 GTF2IP7 POM121C ZP3 |
nsv1143414 | Chr.7:76353392 - 76365602 on Build GRCh38 | Deletion | YWHAG |
nsv1028665 | Chr.7:76336044 - 76445651 on Build GRCh38 | Gain | YWHAG SSC4D ZP3 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | POMZP3 HIP1 MDH2 MIR4651 CCL26 POR STYXL1 SPDYE16 LOC105375355 SSC4D DTX2 SPDYE5 PMS2P3 YWHAG CCL24 RHBDD2 SNORA14A HSPB1 FDPSP2 TMEM120A LOC107986711 LOC100133091 SRRM3 GTF2IP7 POM121C ZP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |