Assay Details
Target Gene Details
Entrez Gene ID: | 7532 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma |
Gene Aliases: |
14-3-3GAMMA, PPP1R170 |
Location: |
Chr.7:76326791-76359025 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YWHAG | NM_012479.3 | NP_036611.2 | ||
AB024334.1 | BAA85184.1 | |||
AF142498.1 | AAD48408.1 | |||
AK024230.1 | ||||
AK293505.1 | ||||
AK295029.1 | ||||
BC020963.2 | AAH20963.1 | |||
CR541904.1 | CAG46702.1 | |||
CR541925.1 | CAG46723.1 | |||
DC343104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422494 | Chr.7:76286900 - 76984449 on Build GRCh38 | Duplication | DTX2 DTX2P1-UPK3BP1-PMS2P11 SRRM3 SSC4D LOC105375355 YWHAG LOC100133091 HSPB1 POMZP3 ZP3 SPDYE16 FDPSP2 |
nsv428174 | Chr.7:76305019 - 77220062 on Build GRCh38 | Gain | DTX2 YWHAG LOC100133091 ZP3 SPDYE16 PMS2P9 SPDYE17 DTX2P1-UPK3BP1-PMS2P11 SSC4D LOC105375355 POMZP3 CCDC146 FGL2 SPDYE18 FDPSP2 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | DTX2 SRRM3 STAG3L1 RHBDD2 PMS2P3 ZP3 SPDYE16 PMS2P2 POR SSC4D SNORA14A GTF2IP1 MIR4651 HSPB1 SPDYE5 MDH2 STYXL1 CCL24 YWHAG LOC100133091 NCF1C LOC541473 POM121C TRIM73 NSUN5P1 LOC107986711 LOC105375355 GTF2IP7 POMZP3 CCL26 HIP1 LOC441259 TMEM120A GTF2IRD2B PMS2P10 FDPSP2 |
nsv1143414 | Chr.7:76353392 - 76365602 on Build GRCh38 | Deletion | YWHAG |
nsv1028665 | Chr.7:76336044 - 76445651 on Build GRCh38 | Gain | SSC4D YWHAG ZP3 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | DTX2 SRRM3 RHBDD2 PMS2P3 ZP3 SPDYE16 POR SSC4D SNORA14A MIR4651 HSPB1 SPDYE5 MDH2 STYXL1 CCL24 YWHAG LOC100133091 POM121C LOC107986711 LOC105375355 GTF2IP7 POMZP3 CCL26 HIP1 TMEM120A FDPSP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |