Assay Details
Target Gene Details
Entrez Gene ID: | 5447 |
Gene Name: | cytochrome p450 oxidoreductase |
Gene Aliases: |
CPR, CYPOR, P450R |
Location: |
Chr.7:75915102-75986855 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POR | NM_000941.2 | NP_000932.3 | ||
AK129978.1 | ||||
AK290529.1 | ||||
AK312543.1 | ||||
BP244929.1 | ||||
BX648619.1 | CAH56151.1 | |||
DA701694.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain | STYXL1 MDH2 LOC107986711 SNORA14A GTF2IP7 TMEM120A POR |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | MIR4651 SPDYE5 MDH2 LOC541473 SSC4D TRIM73 PMS2P2 HIP1 SRRM3 PMS2P10 SNORA14A NSUN5P1 GTF2IP7 CCL26 PMS2P3 LOC100133091 FDPSP2 YWHAG STAG3L1 POMZP3 ZP3 CCL24 RHBDD2 SPDYE16 NCF1C POR LOC105375355 POM121C HSPB1 STYXL1 GTF2IP1 LOC441259 LOC107986711 GTF2IRD2B TMEM120A DTX2 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | MIR4651 STYXL1 SNORA14A TMEM120A POR |
esv3613744 | Chr.7:75937441 - 75957916 on Build GRCh38 | Loss | SNORA14A POR |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | MIR4651 SPDYE5 MDH2 SSC4D HIP1 SRRM3 SNORA14A GTF2IP7 CCL26 PMS2P3 LOC100133091 FDPSP2 YWHAG POMZP3 ZP3 CCL24 RHBDD2 SPDYE16 POR LOC105375355 POM121C HSPB1 STYXL1 LOC107986711 TMEM120A DTX2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |