Assay Details
Target Gene Details
Entrez Gene ID: | 51657 |
Gene Name: | serine/threonine/tyrosine interacting-like 1 |
Gene Aliases: |
DUSP24, MK-STYX, MKSTYX |
Location: |
Chr.7:75995820-76048709 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain | MDH2 GTF2IP7 LOC107986711 POR TMEM120A SNORA14A STYXL1 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | SPDYE5 TRIM73 GTF2IP7 POMZP3 CCL26 PMS2P10 FDPSP2 NSUN5P1 TMEM120A MDH2 MIR4651 LOC441259 POR DTX2 SRRM3 HSPB1 PMS2P2 GTF2IRD2B LOC541473 PMS2P3 LOC105375355 ZP3 POM121C LOC107986711 NCF1C HIP1 STYXL1 YWHAG STAG3L1 RHBDD2 GTF2IP1 LOC100133091 SSC4D SPDYE16 SNORA14A CCL24 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | MIR4651 POR TMEM120A SNORA14A STYXL1 |
esv3613749 | Chr.7:76009631 - 76035709 on Build GRCh38 | Loss | STYXL1 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | SPDYE5 GTF2IP7 POMZP3 CCL26 FDPSP2 TMEM120A MDH2 MIR4651 POR DTX2 SRRM3 HSPB1 PMS2P3 LOC105375355 ZP3 POM121C LOC107986711 HIP1 STYXL1 YWHAG RHBDD2 LOC100133091 SSC4D SPDYE16 SNORA14A CCL24 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |