Assay Details
Target Gene Details
Entrez Gene ID: | 8468 |
Gene Name: | FK506 binding protein 6 |
Gene Aliases: |
FKBP36 |
Location: |
Chr.7:73328152-73358637 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FKBP6 | NM_001135211.2 | NP_001128683.1 | ||
NM_001281304.1 | NP_001268233.1 | |||
NM_003602.4 | NP_003593.3 | |||
XM_006716153.2 | XP_006716216.1 | |||
XM_017012741.1 | XP_016868230.1 | |||
AF038847.1 | AAC64249.1 | |||
AF447060.1 | AAP97324.1 | |||
AK302121.1 | ||||
BC036817.1 | AAH36817.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759537 | Chr.7:72446485 - 73461568 on Build GRCh38 | Gain+Loss | SPDYE7P POM121 SBDSP1 GTF2IP4 NCF1B BAZ1B PMS2P7 TRIM50 GTF2IRD2P1 NSUN5P2 TYW1B CALN1 NSUN5 SPDYE8P PMS2P6 FZD9 LOC100101148 FKBP6 MIR4650-2 TRIM74 SPDYE11 STAG3L3 |
esv3891145 | Chr.7:73308984 - 73364098 on Build GRCh38 | Gain | TRIM50 FKBP6 |
dgv6466n100 | Chr.7:73245562 - 73420814 on Build GRCh38 | Gain | NSUN5 TRIM50 FKBP6 GTF2IRD2P1 |
nsv831027 | Chr.7:73173922 - 73442066 on Build GRCh38 | Loss | NSUN5 GTF2IP4 NCF1B BAZ1B FZD9 TRIM50 FKBP6 GTF2IRD2P1 |
nsv1023935 | Chr.7:73338347 - 73398957 on Build GRCh38 | Gain | FKBP6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |