Assay Details
Target Gene Details
Entrez Gene ID: | 5447 |
Gene Name: | cytochrome p450 oxidoreductase |
Gene Aliases: |
CPR, CYPOR, P450R |
Location: |
Chr.7:75915102-75986855 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POR | NM_000941.2 | NP_000932.3 | ||
AB051763.1 | BAB18572.1 | |||
AB209874.1 | BAD93111.1 | |||
AF258341.1 | AAG09798.1 | |||
AK129978.1 | ||||
AK290529.1 | ||||
AK312543.1 | ||||
BC034277.1 | AAH34277.1 | |||
BP244929.1 | ||||
BX648619.1 | CAH56151.1 | |||
DA701694.1 | ||||
S90469.1 | AAB21814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain | MDH2 SNORA14A POR STYXL1 TMEM120A GTF2IP7 LOC107986711 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | DTX2 POM121C TMEM120A MIR4651 TRIM73 LOC107986711 SPDYE16 HIP1 LOC100133091 POR STYXL1 PMS2P2 ZP3 PMS2P3 CCL26 GTF2IRD2B SRRM3 STAG3L1 CCL24 NCF1C FDPSP2 LOC105375355 GTF2IP7 NSUN5P1 GTF2IP1 LOC441259 YWHAG MDH2 POMZP3 SNORA14A RHBDD2 SSC4D LOC541473 PMS2P10 SPDYE5 HSPB1 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | SNORA14A POR STYXL1 TMEM120A MIR4651 |
esv3613744 | Chr.7:75937441 - 75957916 on Build GRCh38 | Loss | SNORA14A POR |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | DTX2 POM121C TMEM120A MIR4651 LOC107986711 SPDYE16 HIP1 LOC100133091 POR STYXL1 ZP3 PMS2P3 CCL26 SRRM3 CCL24 FDPSP2 LOC105375355 GTF2IP7 YWHAG MDH2 POMZP3 SNORA14A RHBDD2 SSC4D SPDYE5 HSPB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |