Assay Details
Target Gene Details
Entrez Gene ID: | 5447 |
Gene Name: | cytochrome p450 oxidoreductase |
Gene Aliases: |
CPR, CYPOR, P450R |
Location: |
Chr.7:75915102-75986855 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POR | NM_000941.2 | NP_000932.3 | ||
AK129978.1 | ||||
AK290529.1 | ||||
AK312543.1 | ||||
BP244929.1 | ||||
BX648619.1 | CAH56151.1 | |||
DA701694.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv3613744 | Chr.7:75937441 - 75957916 on Build GRCh38 | Loss |
![]() ![]() |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)