Assay Details
Target Gene Details
Entrez Gene ID: | 7532 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma |
Gene Aliases: |
14-3-3GAMMA, PPP1R170 |
Location: |
Chr.7:76326791-76359025 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YWHAG | NM_012479.3 | NP_036611.2 | ||
AB024334.1 | BAA85184.1 | |||
AF142498.1 | AAD48408.1 | |||
AK024230.1 | ||||
AK293505.1 | ||||
AK295029.1 | ||||
BC020963.2 | AAH20963.1 | |||
CR541904.1 | CAG46702.1 | |||
CR541925.1 | CAG46723.1 | |||
DC343104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422494 | Chr.7:76286900 - 76984449 on Build GRCh38 | Duplication | DTX2 ZP3 POMZP3 YWHAG SSC4D SRRM3 SPDYE16 LOC105375355 LOC100133091 FDPSP2 HSPB1 DTX2P1-UPK3BP1-PMS2P11 |
nsv428174 | Chr.7:76305019 - 77220062 on Build GRCh38 | Gain | ZP3 PMS2P9 SSC4D SPDYE16 FGL2 SPDYE18 DTX2P1-UPK3BP1-PMS2P11 CCDC146 DTX2 POMZP3 YWHAG LOC105375355 LOC100133091 SPDYE17 FDPSP2 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | POR PMS2P10 STYXL1 SPDYE16 SPDYE5 GTF2IP1 PMS2P2 RHBDD2 TMEM120A CCL24 POMZP3 LOC100133091 MIR4651 LOC107986711 ZP3 MDH2 SSC4D SRRM3 NCF1C PMS2P3 TRIM73 STAG3L1 NSUN5P1 DTX2 GTF2IRD2B CCL26 GTF2IP7 LOC441259 SNORA14A YWHAG POM121C LOC105375355 LOC541473 FDPSP2 HSPB1 HIP1 |
nsv1028665 | Chr.7:76336044 - 76445651 on Build GRCh38 | Gain | ZP3 YWHAG SSC4D |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | POR STYXL1 SPDYE16 SPDYE5 RHBDD2 TMEM120A CCL24 POMZP3 LOC100133091 MIR4651 LOC107986711 ZP3 MDH2 SSC4D SRRM3 PMS2P3 DTX2 CCL26 GTF2IP7 SNORA14A YWHAG POM121C LOC105375355 FDPSP2 HSPB1 HIP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |