Assay Details
Target Gene Details
Entrez Gene ID: | 9296 |
Gene Name: | ATPase H+ transporting V1 subunit F |
Gene Aliases: |
ATP6S14, VATF, Vma7 |
Location: |
Chr.7:128862803-128865849 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP6V1F | NM_001198909.1 | NP_001185838.1 | ||
NM_004231.3 | NP_004222.2 | |||
AK312214.1 | ||||
BC104230.1 | AAI04231.1 | |||
BC104231.1 | AAI04232.1 | |||
BC107854.1 | AAI07855.1 | |||
BG724326.1 | ||||
BU683155.1 | ||||
BU956696.1 | ||||
CR456896.1 | CAG33177.1 | |||
D49400.1 | BAA08392.1 | |||
DB499437.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv464717 | Chr.7:128832927 - 128876891 on Build GRCh38 | Loss | LOC100130705 ATP6V1F FLNC KCP |
esv2670312 | Chr.7:128499519 - 128999613 on Build GRCh38 | Deletion | TNPO3 CCDC136 FAM71F1 CALU IRF5 LINC01000 LOC100130705 ATP6V1F FAM71F2 FLNC METTL2B OPN1SW KCP |
nsv608409 | Chr.7:128843173 - 128911686 on Build GRCh38 | Loss | LOC100130705 ATP6V1F FLNC KCP |
esv2735137 | Chr.7:128499440 - 128999664 on Build GRCh38 | Deletion | TNPO3 CCDC136 FAM71F1 CALU IRF5 LINC01000 LOC100130705 ATP6V1F FAM71F2 FLNC METTL2B OPN1SW KCP |
dgv11612n54 | Chr.7:128818595 - 128928906 on Build GRCh38 | Loss | LOC100130705 ATP6V1F FLNC CCDC136 KCP |
dgv11611n54 | Chr.7:128794846 - 128881377 on Build GRCh38 | Loss | LOC100130705 ATP6V1F FLNC CCDC136 KCP |
nsv831125 | Chr.7:128772651 - 128955697 on Build GRCh38 | Loss | LOC100130705 TNPO3 ATP6V1F FLNC CCDC136 CALU OPN1SW IRF5 KCP |
esv22837 | Chr.7:128774435 - 128953151 on Build GRCh38 | Gain | LOC100130705 ATP6V1F FLNC CCDC136 OPN1SW IRF5 KCP |
nsv517923 | Chr.7:128805212 - 128876891 on Build GRCh38 | Gain | LOC100130705 ATP6V1F FLNC CCDC136 KCP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |