Assay Details
Target Gene Details
Entrez Gene ID: | 26261 |
Gene Name: | F-box protein 24 |
Gene Aliases: |
FBX24 |
Location: |
Chr.7:100582490-100601117 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FBXO24 | XM_017011961.1 | XP_016867450.1 |
Target Gene Details
Entrez Gene ID: | 4034 |
Gene Name: | leucine rich repeats and calponin homology domain containing 4 |
Gene Aliases: |
LRN, LRRN1, LRRN4, PP14183 |
Location: |
Chr.7:100574011-100586188 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRCH4 | NM_001289934.1 | NP_001276863.1 | ||
NM_002319.4 | NP_002310.2 | |||
AK074833.1 | ||||
AK097420.1 | ||||
AK302410.1 | ||||
BC018529.1 | AAH18529.3 | |||
BG722226.1 | ||||
DC397219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv607932 | Chr.7:100574872 - 100585068 on Build GRCh38 | Loss |
![]() ![]() |
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv951386 | Chr.7:100566478 - 100682977 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv831072 | Chr.7:100564880 - 100765212 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv5868 | Chr.7:100572375 - 100617023 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)