Assay Details
Target Gene Details
Entrez Gene ID: | 7532 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma |
Gene Aliases: |
14-3-3GAMMA, PPP1R170 |
Location: |
Chr.7:76326791-76359025 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YWHAG | NM_012479.3 | NP_036611.2 | ||
AB024334.1 | BAA85184.1 | |||
AF142498.1 | AAD48408.1 | |||
AK024230.1 | ||||
AK293505.1 | ||||
AK295029.1 | ||||
BC020963.2 | AAH20963.1 | |||
CR541904.1 | CAG46702.1 | |||
CR541925.1 | CAG46723.1 | |||
DC343104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422494 | Chr.7:76286900 - 76984449 on Build GRCh38 | Duplication | SSC4D YWHAG ZP3 SPDYE16 SRRM3 FDPSP2 DTX2P1-UPK3BP1-PMS2P11 DTX2 LOC105375355 LOC100133091 HSPB1 POMZP3 |
nsv428174 | Chr.7:76305019 - 77220062 on Build GRCh38 | Gain | YWHAG SPDYE16 FDPSP2 FGL2 LOC105375355 LOC100133091 SSC4D CCDC146 ZP3 DTX2P1-UPK3BP1-PMS2P11 SPDYE17 DTX2 SPDYE18 POMZP3 PMS2P9 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | CCL26 GTF2IP7 YWHAG POM121C FDPSP2 HIP1 NCF1C MIR4651 LOC100133091 POR HSPB1 NSUN5P1 SSC4D ZP3 STAG3L1 LOC541473 TMEM120A GTF2IRD2B PMS2P10 PMS2P2 DTX2 SNORA14A TRIM73 STYXL1 SPDYE16 SRRM3 LOC441259 PMS2P3 GTF2IP1 LOC105375355 LOC107986711 RHBDD2 CCL24 MDH2 POMZP3 SPDYE5 |
nsv1028665 | Chr.7:76336044 - 76445651 on Build GRCh38 | Gain | SSC4D YWHAG ZP3 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | CCL26 GTF2IP7 YWHAG POM121C FDPSP2 HIP1 MIR4651 LOC100133091 POR HSPB1 SSC4D ZP3 TMEM120A DTX2 SNORA14A STYXL1 SPDYE16 SRRM3 PMS2P3 LOC105375355 LOC107986711 RHBDD2 CCL24 MDH2 POMZP3 SPDYE5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |