Assay Details
Target Gene Details
Entrez Gene ID: | 5447 |
Gene Name: | cytochrome p450 oxidoreductase |
Gene Aliases: |
CPR, CYPOR, P450R |
Location: |
Chr.7:75915102-75986855 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POR | NM_000941.2 | NP_000932.3 | ||
AK129978.1 | ||||
AK290529.1 | ||||
AK312543.1 | ||||
BP244929.1 | ||||
BX648619.1 | CAH56151.1 | |||
DA701694.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | POM121C PMS2P2 LOC541473 TMEM120A STAG3L1 LOC105375355 SRRM3 POR SPDYE5 FDPSP2 SSC4D LOC107986711 RHBDD2 POMZP3 NCF1C NSUN5P1 MIR4651 DTX2 LOC100133091 STYXL1 SNORA14A GTF2IP7 CCL24 GTF2IP1 SPDYE16 HSPB1 HIP1 ZP3 YWHAG GTF2IRD2B PMS2P10 PMS2P3 LOC441259 MDH2 TRIM73 CCL26 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | MIR4651 STYXL1 TMEM120A SNORA14A POR |
esv3613743 | Chr.7:75917905 - 75929949 on Build GRCh38 | Loss | POR |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | POM121C TMEM120A LOC105375355 SRRM3 POR SPDYE5 FDPSP2 SSC4D LOC107986711 RHBDD2 POMZP3 MIR4651 DTX2 LOC100133091 STYXL1 SNORA14A GTF2IP7 CCL24 SPDYE16 HSPB1 HIP1 ZP3 YWHAG PMS2P3 MDH2 CCL26 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |