Assay Details
Target Gene Details
Entrez Gene ID: | 51657 |
Gene Name: | serine/threonine/tyrosine interacting-like 1 |
Gene Aliases: |
DUSP24, MK-STYX, MKSTYX |
Location: |
Chr.7:75995820-76048709 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain | LOC107986711 SNORA14A STYXL1 POR MDH2 TMEM120A GTF2IP7 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | YWHAG FDPSP2 SNORA14A HIP1 MDH2 PMS2P3 SSC4D SPDYE5 GTF2IRD2B PMS2P10 STYXL1 NCF1C CCL24 DTX2 LOC541473 TMEM120A HSPB1 GTF2IP7 MIR4651 LOC441259 GTF2IP1 LOC105375355 POMZP3 RHBDD2 SPDYE16 PMS2P2 POR NSUN5P1 CCL26 STAG3L1 POM121C LOC107986711 LOC100133091 SRRM3 TRIM73 ZP3 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | SNORA14A STYXL1 POR TMEM120A MIR4651 |
esv3613749 | Chr.7:76009631 - 76035709 on Build GRCh38 | Loss | STYXL1 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | YWHAG FDPSP2 SNORA14A HIP1 MDH2 PMS2P3 SSC4D SPDYE5 STYXL1 CCL24 DTX2 TMEM120A HSPB1 GTF2IP7 MIR4651 LOC105375355 POMZP3 RHBDD2 SPDYE16 POR CCL26 POM121C LOC107986711 LOC100133091 SRRM3 ZP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |