Assay Details
Target Gene Details
Entrez Gene ID: | 7532 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma |
Gene Aliases: |
14-3-3GAMMA, PPP1R170 |
Location: |
Chr.7:76326791-76359025 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YWHAG | NM_012479.3 | NP_036611.2 | ||
AB024334.1 | BAA85184.1 | |||
AF142498.1 | AAD48408.1 | |||
AK024230.1 | ||||
AK293505.1 | ||||
AK295029.1 | ||||
BC020963.2 | AAH20963.1 | |||
CR541904.1 | CAG46702.1 | |||
CR541925.1 | CAG46723.1 | |||
DC343104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422494 | Chr.7:76286900 - 76984449 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv428174 | Chr.7:76305019 - 77220062 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1028665 | Chr.7:76336044 - 76445651 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)