Assay Details
Target Gene Details
Entrez Gene ID: | 10344 |
Gene Name: | C-C motif chemokine ligand 26 |
Gene Aliases: |
IMAC, MIP-4a, MIP-4alpha, SCYA26, TSC-1 |
Location: |
Chr.7:75769524-75791583 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL26 | NM_006072.4 | NP_006063.1 | ||
XM_017011672.1 | XP_016867161.1 | |||
AY358893.1 | AAQ89252.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3613737 | Chr.7:75769270 - 75786583 on Build GRCh38 | Gain | CCL26 |
dgv6468n100 | Chr.7:75701432 - 75777782 on Build GRCh38 | Gain | CCL26 HIP1 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | GTF2IRD2B TRIM73 PMS2P2 LOC105375355 HIP1 NSUN5P1 SRRM3 POR POMZP3 HSPB1 YWHAG MIR4651 RHBDD2 CCL24 POM121C FDPSP2 PMS2P3 SSC4D STYXL1 LOC100133091 LOC441259 STAG3L1 GTF2IP7 LOC107986711 ZP3 PMS2P10 CCL26 SPDYE16 LOC541473 GTF2IP1 TMEM120A SNORA14A SPDYE5 MDH2 DTX2 NCF1C |
nsv518028 | Chr.7:75539242 - 75825134 on Build GRCh38 | Gain | CCL24 CCL26 HIP1 |
esv3613726 | Chr.7:75699055 - 75796413 on Build GRCh38 | Gain | CCL26 HIP1 |
esv2764051 | Chr.7:75663839 - 75875518 on Build GRCh38 | Gain+Loss | CCL24 CCL26 HIP1 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | LOC105375355 HIP1 SRRM3 POR POMZP3 HSPB1 YWHAG MIR4651 RHBDD2 CCL24 POM121C FDPSP2 PMS2P3 SSC4D STYXL1 LOC100133091 GTF2IP7 LOC107986711 ZP3 CCL26 SPDYE16 TMEM120A SNORA14A SPDYE5 MDH2 DTX2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |