Assay Details
Target Gene Details
Entrez Gene ID: | 7532 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma |
Gene Aliases: |
14-3-3GAMMA, PPP1R170 |
Location: |
Chr.7:76326791-76359025 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YWHAG | NM_012479.3 | NP_036611.2 | ||
AB024334.1 | BAA85184.1 | |||
AF142498.1 | AAD48408.1 | |||
AK024230.1 | ||||
AK293505.1 | ||||
AK295029.1 | ||||
BC020963.2 | AAH20963.1 | |||
CR541904.1 | CAG46702.1 | |||
CR541925.1 | CAG46723.1 | |||
DC343104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422494 | Chr.7:76286900 - 76984449 on Build GRCh38 | Duplication | DTX2 DTX2P1-UPK3BP1-PMS2P11 FDPSP2 SSC4D ZP3 SRRM3 POMZP3 LOC105375355 LOC100133091 SPDYE16 YWHAG HSPB1 |
nsv428174 | Chr.7:76305019 - 77220062 on Build GRCh38 | Gain | FGL2 DTX2P1-UPK3BP1-PMS2P11 POMZP3 PMS2P9 SPDYE16 SPDYE18 DTX2 FDPSP2 SSC4D SPDYE17 ZP3 LOC105375355 CCDC146 LOC100133091 YWHAG |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | TMEM120A GTF2IRD2B POMZP3 POM121C SNORA14A POR HIP1 STYXL1 NSUN5P1 SPDYE16 LOC541473 FDPSP2 CCL26 SPDYE5 CCL24 GTF2IP7 GTF2IP1 NCF1C STAG3L1 LOC100133091 MDH2 HSPB1 PMS2P2 TRIM73 PMS2P10 RHBDD2 SRRM3 MIR4651 LOC107986711 LOC441259 DTX2 SSC4D ZP3 PMS2P3 LOC105375355 YWHAG |
nsv1028665 | Chr.7:76336044 - 76445651 on Build GRCh38 | Gain | SSC4D ZP3 YWHAG |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | TMEM120A POMZP3 POM121C SNORA14A POR HIP1 STYXL1 SPDYE16 FDPSP2 CCL26 SPDYE5 CCL24 GTF2IP7 LOC100133091 MDH2 HSPB1 RHBDD2 SRRM3 MIR4651 LOC107986711 DTX2 SSC4D ZP3 PMS2P3 LOC105375355 YWHAG |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |