Assay Details
Target Gene Details
Entrez Gene ID: | 7532 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma |
Gene Aliases: |
14-3-3GAMMA, PPP1R170 |
Location: |
Chr.7:76326791-76359025 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YWHAG | NM_012479.3 | NP_036611.2 | ||
AB024334.1 | BAA85184.1 | |||
AF142498.1 | AAD48408.1 | |||
AK024230.1 | ||||
AK293505.1 | ||||
AK295029.1 | ||||
BC020963.2 | AAH20963.1 | |||
CR541904.1 | CAG46702.1 | |||
CR541925.1 | CAG46723.1 | |||
DC343104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422494 | Chr.7:76286900 - 76984449 on Build GRCh38 | Duplication | HSPB1 SRRM3 DTX2 ZP3 LOC100133091 YWHAG LOC105375355 POMZP3 SPDYE16 SSC4D DTX2P1-UPK3BP1-PMS2P11 FDPSP2 |
nsv428174 | Chr.7:76305019 - 77220062 on Build GRCh38 | Gain | DTX2 LOC100133091 SPDYE17 PMS2P9 FGL2 YWHAG LOC105375355 POMZP3 DTX2P1-UPK3BP1-PMS2P11 CCDC146 ZP3 SPDYE16 SPDYE18 SSC4D FDPSP2 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | SPDYE5 SRRM3 DTX2 STAG3L1 LOC105375355 SNORA14A NCF1C PMS2P10 NSUN5P1 POM121C CCL26 GTF2IP7 TRIM73 MDH2 HIP1 FDPSP2 PMS2P3 GTF2IRD2B HSPB1 CCL24 POR TMEM120A LOC100133091 LOC541473 YWHAG POMZP3 LOC107986711 RHBDD2 LOC441259 ZP3 MIR4651 STYXL1 PMS2P2 GTF2IP1 SPDYE16 SSC4D |
nsv1028665 | Chr.7:76336044 - 76445651 on Build GRCh38 | Gain | ZP3 YWHAG SSC4D |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | SPDYE5 SRRM3 DTX2 LOC105375355 SNORA14A POM121C CCL26 GTF2IP7 MDH2 HIP1 FDPSP2 PMS2P3 HSPB1 CCL24 POR TMEM120A LOC100133091 YWHAG POMZP3 LOC107986711 RHBDD2 ZP3 MIR4651 STYXL1 SPDYE16 SSC4D |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |