Assay Details
Target Gene Details
Entrez Gene ID: | 5447 |
Gene Name: | cytochrome p450 oxidoreductase |
Gene Aliases: |
CPR, CYPOR, P450R |
Location: |
Chr.7:75915102-75986855 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POR | NM_000941.2 | NP_000932.3 | ||
AK129978.1 | ||||
AK290529.1 | ||||
AK312543.1 | ||||
BP244929.1 | ||||
BX648619.1 | CAH56151.1 | |||
DA701694.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | SPDYE16 DTX2 LOC541473 PMS2P3 LOC107986711 FDPSP2 TRIM73 PMS2P2 LOC441259 YWHAG SPDYE5 ZP3 NSUN5P1 NCF1C STAG3L1 TMEM120A LOC100133091 SNORA14A POR RHBDD2 SRRM3 CCL26 PMS2P10 MIR4651 POM121C GTF2IP1 LOC105375355 MDH2 POMZP3 STYXL1 HIP1 GTF2IP7 GTF2IRD2B CCL24 SSC4D HSPB1 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | STYXL1 MIR4651 TMEM120A SNORA14A POR |
esv3613743 | Chr.7:75917905 - 75929949 on Build GRCh38 | Loss | POR |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | SPDYE16 DTX2 PMS2P3 LOC107986711 FDPSP2 YWHAG SPDYE5 ZP3 TMEM120A LOC100133091 SNORA14A POR RHBDD2 SRRM3 CCL26 MIR4651 POM121C LOC105375355 MDH2 POMZP3 STYXL1 HIP1 GTF2IP7 CCL24 SSC4D HSPB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |