Assay Details
Target Gene Details
Entrez Gene ID: | 10268 |
Gene Name: | receptor (G protein-coupled) activity modifying protein 3 |
Gene Aliases: |
- |
Location: |
Chr.7:45157768-45184250 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAMP3 | NM_005856.2 | NP_005847.1 | ||
XM_006715631.3 | XP_006715694.1 | |||
AJ001016.1 | CAA04474.1 | |||
AK222469.1 | BAD96189.1 | |||
AK311871.1 | ||||
AY265459.1 | AAP23300.1 | |||
BC022304.1 | AAH22304.1 | |||
BC053852.1 | AAH53852.1 | |||
BP347072.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv516731 | Chr.7:45172191 - 45185503 on Build GRCh38 | Loss |
![]() |
nsv830986 | Chr.7:45129807 - 45319056 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)