Assay Details
Target Gene Details
Entrez Gene ID: | 7532 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma |
Gene Aliases: |
14-3-3GAMMA, PPP1R170 |
Location: |
Chr.7:76326791-76359025 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YWHAG | NM_012479.3 | NP_036611.2 | ||
AB024334.1 | BAA85184.1 | |||
AF142498.1 | AAD48408.1 | |||
AK024230.1 | ||||
AK293505.1 | ||||
AK295029.1 | ||||
BC020963.2 | AAH20963.1 | |||
CR541904.1 | CAG46702.1 | |||
CR541925.1 | CAG46723.1 | |||
DC343104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422494 | Chr.7:76286900 - 76984449 on Build GRCh38 | Duplication | LOC105375355 SPDYE16 LOC100133091 SSC4D HSPB1 YWHAG SRRM3 DTX2 ZP3 POMZP3 DTX2P1-UPK3BP1-PMS2P11 FDPSP2 |
nsv428174 | Chr.7:76305019 - 77220062 on Build GRCh38 | Gain | SPDYE16 SPDYE17 SSC4D POMZP3 CCDC146 DTX2P1-UPK3BP1-PMS2P11 FDPSP2 LOC105375355 LOC100133091 YWHAG SPDYE18 DTX2 ZP3 PMS2P9 FGL2 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | GTF2IP7 SPDYE16 SPDYE5 CCL24 PMS2P2 HSPB1 TMEM120A POM121C PMS2P3 FDPSP2 MIR4651 RHBDD2 LOC105375355 PMS2P10 LOC100133091 NCF1C CCL26 HIP1 SRRM3 MDH2 LOC441259 GTF2IP1 LOC107986711 GTF2IRD2B TRIM73 SSC4D STYXL1 LOC541473 POR STAG3L1 POMZP3 SNORA14A YWHAG NSUN5P1 DTX2 ZP3 |
nsv1028665 | Chr.7:76336044 - 76445651 on Build GRCh38 | Gain | SSC4D YWHAG ZP3 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | GTF2IP7 SPDYE16 SPDYE5 CCL24 HSPB1 TMEM120A POM121C PMS2P3 FDPSP2 MIR4651 RHBDD2 LOC105375355 LOC100133091 CCL26 HIP1 SRRM3 MDH2 LOC107986711 SSC4D STYXL1 POR POMZP3 SNORA14A YWHAG DTX2 ZP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |