Assay Details
Target Gene Details
Entrez Gene ID: | 5447 |
Gene Name: | cytochrome p450 oxidoreductase |
Gene Aliases: |
CPR, CYPOR, P450R |
Location: |
Chr.7:75915102-75986855 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POR | NM_000941.2 | NP_000932.3 | ||
AB051763.1 | BAB18572.1 | |||
AB209874.1 | BAD93111.1 | |||
AF258341.1 | AAG09798.1 | |||
AK129978.1 | ||||
AK290529.1 | ||||
AK312543.1 | ||||
BC034277.1 | AAH34277.1 | |||
BP244929.1 | ||||
BX648619.1 | CAH56151.1 | |||
DA701694.1 | ||||
S90469.1 | AAB21814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain | MDH2 LOC107986711 TMEM120A STYXL1 SNORA14A POR GTF2IP7 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | MDH2 YWHAG STAG3L1 RHBDD2 STYXL1 SNORA14A SSC4D LOC100133091 HSPB1 SPDYE16 TMEM120A PMS2P2 CCL26 NCF1C LOC441259 POMZP3 PMS2P10 NSUN5P1 GTF2IP1 LOC107986711 HIP1 FDPSP2 TRIM73 LOC541473 ZP3 MIR4651 PMS2P3 DTX2 SRRM3 GTF2IP7 LOC105375355 GTF2IRD2B POM121C CCL24 SPDYE5 POR |
esv3613746 | Chr.7:75965095 - 76003876 on Build GRCh38 | Loss | TMEM120A STYXL1 POR |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | TMEM120A STYXL1 SNORA14A MIR4651 POR |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | MDH2 YWHAG RHBDD2 STYXL1 SNORA14A SSC4D LOC100133091 HSPB1 SPDYE16 TMEM120A CCL26 POMZP3 LOC107986711 HIP1 FDPSP2 ZP3 MIR4651 PMS2P3 DTX2 SRRM3 GTF2IP7 LOC105375355 POM121C CCL24 SPDYE5 POR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |