Assay Details
Target Gene Details
Entrez Gene ID: | 51657 |
Gene Name: | serine/threonine/tyrosine interacting-like 1 |
Gene Aliases: |
DUSP24, MK-STYX, MKSTYX |
Location: |
Chr.7:75995820-76048709 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3613751 | Chr.7:76032280 - 76033713 on Build GRCh38 | Loss | STYXL1 |
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain | MDH2 STYXL1 POR LOC107986711 SNORA14A TMEM120A GTF2IP7 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | LOC441259 SPDYE5 SRRM3 LOC105375355 CCL24 PMS2P10 TMEM120A YWHAG HSPB1 NSUN5P1 POM121C POR SNORA14A SSC4D SPDYE16 STAG3L1 POMZP3 DTX2 PMS2P2 MDH2 LOC100133091 GTF2IRD2B LOC541473 STYXL1 GTF2IP1 LOC107986711 MIR4651 RHBDD2 ZP3 TRIM73 FDPSP2 NCF1C PMS2P3 HIP1 CCL26 GTF2IP7 |
esv3613750 | Chr.7:76031380 - 76045732 on Build GRCh38 | Loss | STYXL1 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | STYXL1 POR SNORA14A MIR4651 TMEM120A |
esv3613749 | Chr.7:76009631 - 76035709 on Build GRCh38 | Loss | STYXL1 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | SPDYE5 SRRM3 LOC105375355 CCL24 TMEM120A YWHAG HSPB1 POM121C POR SNORA14A SSC4D SPDYE16 POMZP3 DTX2 MDH2 LOC100133091 STYXL1 LOC107986711 MIR4651 RHBDD2 ZP3 FDPSP2 PMS2P3 HIP1 CCL26 GTF2IP7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |