Assay Details
Target Gene Details
Entrez Gene ID: | 51657 |
Gene Name: | serine/threonine/tyrosine interacting-like 1 |
Gene Aliases: |
DUSP24, MK-STYX, MKSTYX |
Location: |
Chr.7:75995820-76048709 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain | LOC107986711 GTF2IP7 TMEM120A STYXL1 POR MDH2 SNORA14A |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | LOC107986711 SSC4D ZP3 LOC100133091 YWHAG FDPSP2 TMEM120A NSUN5P1 POMZP3 MDH2 LOC541473 SNORA14A GTF2IP7 STAG3L1 PMS2P2 LOC105375355 CCL26 GTF2IP1 GTF2IRD2B DTX2 MIR4651 PMS2P10 STYXL1 RHBDD2 SPDYE5 SPDYE16 HIP1 NCF1C HSPB1 LOC441259 POR POM121C SRRM3 TRIM73 CCL24 PMS2P3 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | MIR4651 TMEM120A STYXL1 POR SNORA14A |
esv3613749 | Chr.7:76009631 - 76035709 on Build GRCh38 | Loss | STYXL1 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | LOC107986711 SSC4D ZP3 LOC100133091 YWHAG FDPSP2 TMEM120A POMZP3 MDH2 SNORA14A GTF2IP7 LOC105375355 CCL26 DTX2 MIR4651 STYXL1 RHBDD2 SPDYE5 SPDYE16 HIP1 HSPB1 POR POM121C SRRM3 CCL24 PMS2P3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |