Assay Details
Target Gene Details
Entrez Gene ID: | 51657 |
Gene Name: | serine/threonine/tyrosine interacting-like 1 |
Gene Aliases: |
DUSP24, MK-STYX, MKSTYX |
Location: |
Chr.7:75995820-76048709 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428173 | Chr.7:75934531 - 76173362 on Build GRCh38 | Gain | LOC107986711 STYXL1 MDH2 POR SNORA14A GTF2IP7 TMEM120A |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | NSUN5P1 SSC4D POM121C SPDYE16 MIR4651 GTF2IP7 LOC100133091 HSPB1 FDPSP2 SPDYE5 CCL26 SRRM3 MDH2 SNORA14A GTF2IRD2B TMEM120A PMS2P3 LOC105375355 STAG3L1 LOC541473 LOC441259 TRIM73 STYXL1 POMZP3 POR RHBDD2 ZP3 NCF1C YWHAG CCL24 LOC107986711 PMS2P2 GTF2IP1 HIP1 PMS2P10 DTX2 |
esv3613742 | Chr.7:75912784 - 76036530 on Build GRCh38 | Loss | MIR4651 STYXL1 POR SNORA14A TMEM120A |
esv3613749 | Chr.7:76009631 - 76035709 on Build GRCh38 | Loss | STYXL1 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | SSC4D POM121C SPDYE16 MIR4651 GTF2IP7 LOC100133091 HSPB1 FDPSP2 SPDYE5 CCL26 SRRM3 MDH2 SNORA14A TMEM120A PMS2P3 LOC105375355 STYXL1 POMZP3 POR RHBDD2 ZP3 YWHAG CCL24 LOC107986711 HIP1 DTX2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |