Assay Details
Target Gene Details
Entrez Gene ID: | 135138 |
Gene Name: | PARK2 coregulated |
Gene Aliases: |
GLUP, HAK005771, PARK2CRG |
Location: |
Chr.6:162726870-163315492 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv605159 | Chr.6:162379124 - 163058475 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv2759489 | Chr.6:162419890 - 162812229 on Build GRCh38 | Gain |
![]() ![]() |
nsv605181 | Chr.6:162738155 - 162800760 on Build GRCh38 | Gain |
![]() |
nsv1015507 | Chr.6:162444113 - 162913747 on Build GRCh38 | Gain |
![]() ![]() |
nsv605180 | Chr.6:162673891 - 162791267 on Build GRCh38 | Loss |
![]() ![]() |
nsv830857 | Chr.6:162643779 - 162824781 on Build GRCh38 | Loss |
![]() ![]() |
esv3571505 | Chr.6:161937230 - 162847872 on Build GRCh38 | Loss |
![]() ![]() |
nsv1026490 | Chr.6:162712401 - 163284676 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
dgv6176n100 | Chr.6:162161146 - 162762531 on Build GRCh38 | Gain |
![]() ![]() |
nsv1018573 | Chr.6:162604329 - 162751777 on Build GRCh38 | Loss |
![]() ![]() |
nsv517562 | Chr.6:162060428 - 162828613 on Build GRCh38 | Gain+Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)