Assay Details
Target Gene Details
Entrez Gene ID: | 4224 |
Gene Name: | meprin A subunit alpha |
Gene Aliases: |
PPHA |
Location: |
Chr.6:46793357-46845987 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEP1A | NM_005588.2 | NP_005579.2 | ||
XM_011514628.1 | XP_011512930.1 | |||
XM_011514629.1 | XP_011512931.1 | |||
AF478685.1 | AAL85339.1 | |||
AK225212.1 | ||||
AK290282.1 | ||||
AK315246.1 | ||||
AK315840.1 | ||||
BC131714.1 | ||||
BC143651.1 | ||||
DA432559.1 | ||||
M82962.1 | AAA21338.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830654 | Chr.6:46755161 - 46938169 on Build GRCh38 | Gain | ADGRF5 ANKRD66 MEP1A LOC101926962 |
esv2759425 | Chr.6:46732445 - 46960334 on Build GRCh38 | Gain+Loss | PLA2G7 ADGRF5 ANKRD66 MEP1A LOC101926962 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |