Assay Details
Target Gene Details
Entrez Gene ID: | 5167 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 1 |
Gene Aliases: |
ARHR2, COLED, M6S1, NPP1, NPPS, PC-1, PCA1, PDNP1 |
Location: |
Chr.6:131808016-131895155 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENPP1 | NM_006208.2 | NP_006199.2 | ||
AK314870.1 | ||||
BC059375.1 | ||||
D12485.1 | BAA02054.1 | |||
M57736.1 | AAA63237.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3610819 | Chr.6:131754426 - 131844936 on Build GRCh38 | Gain | ENPP1 MIR548H5 |
nsv7968 | Chr.6:131596032 - 131999959 on Build GRCh38 | Loss | ENPP1 MED23 MIR548H5 CTAGE9 ENPP3 CTGF OR2A4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |