Assay Details
Target Gene Details
Entrez Gene ID: | 100129461 |
Gene Name: | LYRM4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:5003816-5240496 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LYRM4-AS1 | NR_126015.1 | |||
NR_126016.1 | ||||
NR_126017.1 | ||||
AK094934.1 | ||||
BE788607.1 | ||||
CR736828.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv462622 | Chr.6:4904146 - 5171775 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv1033905 | Chr.6:4955592 - 5080670 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv600872 | Chr.6:4975079 - 5341957 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv1017380 | Chr.6:5007636 - 5152317 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv830577 | Chr.6:4834333 - 5050310 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv2422499 | Chr.6:4745194 - 5072988 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)