Assay Details
Target Gene Details
Entrez Gene ID: | 6648 |
Gene Name: | superoxide dismutase 2, mitochondrial |
Gene Aliases: |
IPO-B, IPOB, MNSOD, MVCD6, Mn-SOD |
Location: |
Chr.6:159679064-159762529 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SOD2 | NM_001322817.1 | NP_001309746.1 | ||
NM_001322819.1 | NP_001309748.1 | |||
NM_001322820.1 | NP_001309749.1 | |||
AK097395.1 | ||||
BP289103.1 | ||||
DA094180.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759483 | Chr.6:159582799 - 159794119 on Build GRCh38 | Gain | SNORA29 MRPL18 SOD2 LOC401281 WTAP LOC100129518 SNORA20 TCP1 ACAT2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |