Assay Details
Target Gene Details
Entrez Gene ID: | 101926934 |
Gene Name: | uncharacterized LOC101926934 |
Gene Aliases: |
- |
Location: |
Chr.6:46670444-46688165 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101926934 | NR_134642.1 | |||
NR_134643.1 | ||||
NR_134644.1 | ||||
AA725776.1 | ||||
AI638322.1 | ||||
AI698692.1 | ||||
BX114358.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)