Assay Details
Target Gene Details
Entrez Gene ID: | 6992 |
Gene Name: | protein phosphatase 1 regulatory inhibitor subunit 11 |
Gene Aliases: |
CFAP255, HCG-V, HCGV, IPP3, TCTE5, TCTEX5 |
Location: |
Chr.6:30066859-30070333 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP1R11 | XM_006715174.3 | 1 | 77 | XP_006715237.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1074389 | Chr.6:29116522 - 31446623 on Build GRCh38 | Deletion | TRIM26 TRIM39 HCG4 MOG ATAT1 OR12D2 PSORS1C3 LINC00243 MUC21 VARS2 PSORS1C2 NRM OR12D3 HLA-C FLOT1 TCF19 PPP1R11 LOC554223 IFITM4P HCG9 MIR877 TUBB TRIM40 ZNRD1ASP TRIM31 UBD SNORD32B HLA-G OR10C1 HLA-J LOC101929006 MUC22 HLA-B GTF2H4 OR11A1 MDC1-AS1 HLA-A OR2H2 CCHCR1 TRIM15 MDC1 IER3 ZNRD1 HCG27 OR5V1 PPP1R10 DHX16 HLA-F-AS1 DPCR1 PPP1R18 MRPS18B MAS1L TRIM39-RPP21 HCG17 SFTA2 RPP21 HCG8 MICA CDSN POU5F1 HCG20 C6orf15 C6orf136 OR2H1 HCG22 MIR6891 HCG4B HLA-H HLA-L ABCF1 HCG18 RNF39 DDR1 LINC01015 PRR3 HLA-K PSORS1C1 OR2J2 OR14J1 MIR4640 LOC105375014 TRIM31-AS1 HLA-F HLA-E GABBR1 TRIM10 GNL1 ZFP57 |
nsv1122126 | Chr.6:28628023 - 31244023 on Build GRCh38 | Deletion | TRIM26 TRIM39 HCG4 MOG ATAT1 OR12D2 PSORS1C3 LINC00243 MUC21 VARS2 LOC100129636 PSORS1C2 NRM TRIM27 OR12D3 FLOT1 TCF19 PPP1R11 LOC554223 IFITM4P HCG9 MIR877 TUBB TRIM40 ZNRD1ASP OR2J3 TRIM31 UBD SNORD32B HLA-G OR10C1 HLA-J LOC101929006 MUC22 GTF2H4 OR11A1 MDC1-AS1 HLA-A OR2H2 CCHCR1 LINC01623 OR2B3 TRIM15 MDC1 OR2W1 IER3 ZNRD1 HCG27 OR5V1 HCG14 PPP1R10 DHX16 HLA-F-AS1 DPCR1 PPP1R18 MRPS18B MAS1L TRIM39-RPP21 HCG17 SFTA2 RPP21 HCG8 CDSN POU5F1 HCG20 LINC01556 C6orf15 C6orf136 OR2H1 HCG22 LINC00533 HCG4B HLA-H HLA-L HCG16 ABCF1 HCG18 RNF39 DDR1 LINC01015 PRR3 HLA-K PSORS1C1 OR2J2 OR14J1 ZNF311 MIR4640 LOC105375014 TRIM31-AS1 HLA-F HLA-E GABBR1 TRIM10 GNL1 ZFP57 |
nsv1144137 | Chr.6:29720323 - 31338123 on Build GRCh38 | Deletion | TRIM26 TRIM39 HCG4 ATAT1 PSORS1C3 LINC00243 MUC21 VARS2 PSORS1C2 NRM HLA-C FLOT1 TCF19 PPP1R11 LOC554223 IFITM4P HCG9 MIR877 TUBB TRIM40 ZNRD1ASP TRIM31 HLA-G HLA-J MUC22 GTF2H4 MDC1-AS1 HLA-A CCHCR1 TRIM15 MDC1 IER3 ZNRD1 HCG27 PPP1R10 DHX16 HLA-F-AS1 DPCR1 PPP1R18 MRPS18B TRIM39-RPP21 HCG17 SFTA2 RPP21 HCG8 CDSN POU5F1 HCG20 C6orf15 C6orf136 HCG22 HCG4B HLA-H HLA-L ABCF1 HCG18 RNF39 DDR1 PRR3 HLA-K PSORS1C1 MIR4640 LOC105375014 TRIM31-AS1 HLA-F HLA-E TRIM10 GNL1 |
esv2731777 | Chr.6:29943636 - 30260756 on Build GRCh38 | Deletion | TRIM40 TRIM26 HLA-L ZNRD1ASP RNF39 TRIM31 HLA-J HCG17 HCG8 TRIM31-AS1 HLA-A TRIM15 TRIM10 ZNRD1 PPP1R11 HCG9 |
nsv601264 | Chr.6:29883666 - 31320716 on Build GRCh38 | Loss | TRIM26 TRIM39 ATAT1 PSORS1C3 LINC00243 MUC21 VARS2 PSORS1C2 NRM HLA-C FLOT1 TCF19 PPP1R11 HCG9 MIR877 TUBB TRIM40 ZNRD1ASP TRIM31 HLA-J MUC22 GTF2H4 MDC1-AS1 HLA-A CCHCR1 TRIM15 MDC1 IER3 ZNRD1 HCG27 PPP1R10 DHX16 DPCR1 PPP1R18 MRPS18B TRIM39-RPP21 HCG17 SFTA2 RPP21 HCG8 CDSN POU5F1 HCG20 C6orf15 C6orf136 HCG22 HCG4B HLA-H HLA-L ABCF1 HCG18 RNF39 DDR1 PRR3 HLA-K PSORS1C1 MIR4640 LOC105375014 TRIM31-AS1 HLA-E TRIM10 GNL1 |
esv3411275 | Chr.6:29981607 - 31490616 on Build GRCh38 | Duplication | TRIM26 TRIM39 ATAT1 HCG26 PSORS1C3 LINC00243 MUC21 VARS2 PSORS1C2 NRM HLA-C FLOT1 TCF19 LOC102725068 PPP1R11 MIR877 TUBB TRIM40 ZNRD1ASP TRIM31 HLA-J MUC22 HLA-B GTF2H4 MDC1-AS1 CCHCR1 TRIM15 MDC1 IER3 ZNRD1 HCG27 PPP1R10 DHX16 DPCR1 PPP1R18 MRPS18B TRIM39-RPP21 HCG17 SFTA2 RPP21 HCG8 MICA HCP5 CDSN POU5F1 HCG20 C6orf15 C6orf136 HCG22 MIR6891 HLA-L ABCF1 HCG18 RNF39 DDR1 PRR3 PSORS1C1 MIR4640 LOC105375014 TRIM31-AS1 HLA-E TRIM10 GNL1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |