Assay Details
Target Gene Details
Entrez Gene ID: | 401253 |
Gene Name: | long intergenic non-protein coding RNA 336 |
Gene Aliases: |
C6orf227, NCRNA00336 |
Location: |
Chr.6:33586106-33593338 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC00336 | NR_027908.1 | |||
AK125740.1 | BAC86269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3608630 | Chr.6:33439052 - 34076935 on Build GRCh38 | Gain | ITPR3 GRM4 LINC00336 LEMD2 SYNGAP1 IP6K3 ZBTB9 MIR7159 LINC01016 GGNBP1 LOC105375026 BAK1 LOC101929188 MIR1275 MLN MIR3934 UQCC2 |
dgv85n68 | Chr.6:33583425 - 33743569 on Build GRCh38 | Loss | ITPR3 LINC00336 LOC101929188 IP6K3 MIR3934 UQCC2 GGNBP1 |
nsv602833 | Chr.6:33567103 - 33620370 on Build GRCh38 | Loss | LINC00336 BAK1 GGNBP1 |