Assay Details
Target Gene Details
Entrez Gene ID: | 8878 |
Gene Name: | sequestosome 1 |
Gene Aliases: |
A170, FTDALS3, OSIL, PDB3, ZIP3, p60, p62, p62B |
Location: |
Chr.5:179806388-179838078 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SQSTM1 | NM_001142298.1 | NP_001135770.1 | ||
NM_001142299.1 | NP_001135771.1 | |||
AK226167.1 | ||||
BC001874.1 | AAH01874.1 | |||
BI549805.1 | ||||
DA541885.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469579 | Chr.5:179757830 - 179933191 on Build GRCh38 | Loss | MGAT4B SQSTM1 LOC100996419 TBC1D9B MAML1 RNF130 LTC4S MIR1229 C5orf45 |
nsv823361 | Chr.5:179786323 - 179824243 on Build GRCh38 | Loss | MGAT4B SQSTM1 LTC4S MIR1229 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |