Assay Details
Target Gene Details
Entrez Gene ID: | 9319 |
Gene Name: | thyroid hormone receptor interactor 13 |
Gene Aliases: |
16E1BP |
Location: |
Chr.5:892854-919348 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TRIP13 | NM_001166260.1 | NP_001159732.1 | ||
NM_004237.3 | NP_004228.1 | |||
XM_011514163.2 | XP_011512465.1 | |||
AK310376.1 | ||||
BC000404.2 | AAH00404.1 | |||
BC019294.1 | AAH19294.1 | |||
BG722761.1 | ||||
CR456744.1 | CAG33025.1 | |||
DB443523.1 | ||||
L40384.1 | AAC41732.1 | |||
U96131.1 | AAB64095.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830186 | Chr.5:845005 - 966855 on Build GRCh38 | Gain | BRD9 ZDHHC11 TRIP13 |
dgv198e55 | Chr.5:685389 - 919382 on Build GRCh38 | Gain | BRD9 ZDHHC11B ZDHHC11 TRIP13 TPPP |
esv2759316 | Chr.5:620048 - 1161361 on Build GRCh38 | Gain+Loss | CEP72 BRD9 ZDHHC11B ZDHHC11 LOC101929895 TRIP13 NKD2 LOC100506688 SLC12A7 MIR4635 TPPP |
nsv596629 | Chr.5:457146 - 1468172 on Build GRCh38 | Gain | CLPTM1L SLC9A3 PP7080 ZDHHC11 SLC12A7 LOC100996325 EXOC3 MIR4635 TPPP CEP72 BRD9 LPCAT1 ZDHHC11B TERT LOC100506688 SLC6A3 SLC6A19 LOC101929895 LINC01511 TRIP13 NKD2 MIR4456 CTD-3080P12.3 MIR4457 LOC100288152 SLC6A18 |
nsv10654 | Chr.5:676170 - 1082573 on Build GRCh38 | Gain+Loss | BRD9 ZDHHC11B ZDHHC11 TRIP13 NKD2 LOC100506688 SLC12A7 MIR4635 TPPP |
nsv509043 | Chr.5:747066 - 915204 on Build GRCh38 | Insertion | BRD9 ZDHHC11B ZDHHC11 TRIP13 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |