Assay Details
Target Gene Details
Entrez Gene ID: | 5229 |
Gene Name: | protein geranylgeranyltransferase type I subunit beta |
Gene Aliases: |
BGGI, GGTI |
Location: |
Chr.5:115200247-115262887 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PGGT1B | NM_005023.3 | NP_005014.2 | ||
XM_005272020.3 | XP_005272077.2 | |||
XM_011543490.2 | XP_011541792.1 | |||
XM_011543492.2 | XP_011541794.1 | |||
AL520655.1 | ||||
AY780790.1 | AAV98360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830454 | Chr.5:115118167 - 115271462 on Build GRCh38 | Gain+Loss | PGGT1B CCDC112 TRIM36 |
nsv1029371 | Chr.5:115205731 - 115255194 on Build GRCh38 | Loss | PGGT1B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |