Assay Details
Target Gene Details
Entrez Gene ID: | 100652758 |
Gene Name: | uncharacterized LOC100652758 |
Gene Aliases: |
- |
Location: |
Chr.5:151769793-151772176 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100652758 | NR_132345.1 | 2 | 1064 | |
NR_132346.1 | 2 | 785 | ||
AB593120.1 | 2 | 700 |