Assay Details
Target Gene Details
Entrez Gene ID: | 441094 |
Gene Name: | NR2F1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:93409356-93581320 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NR2F1-AS1 | NR_021491.2 | 6 | 2587 | |
NR_109820.1 | 5 | 2548 | ||
NR_109821.1 | 7 | 2708 | ||
NR_109822.1 | 3 | 2998 | ||
NR_109823.1 | 5 | 2545 | ||
NR_109824.1 | 6 | 2644 | ||
NR_109825.1 | 9 | 3510 | ||
AK124699.1 | 5 | 3273 | ||
AW194934.1 | 1 | 112 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830402 | Chr.5:93368195 - 93511639 on Build GRCh38 | Gain | NR2F1-AS1 |