Assay Details
Target Gene Details
Entrez Gene ID: | 51149 |
Gene Name: | chromosome 5 open reading frame 45 |
Gene Aliases: |
- |
Location: |
Chr.5:179837266-179858840 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C5orf45 | NM_001017987.2 | NP_001017987.1 | ||
NM_016175.3 | NP_057259.2 | |||
AK293901.1 | ||||
AK295104.1 | ||||
AK298954.1 | ||||
BC069051.1 | AAH69051.1 | |||
BF965313.1 | ||||
BQ642639.1 | ||||
BX537968.1 | CAD97932.1 | |||
BX647375.1 | ||||
CN363239.1 | ||||
CR936785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469579 | Chr.5:179757830 - 179933191 on Build GRCh38 | Loss | LOC100996419 C5orf45 MGAT4B MAML1 LTC4S TBC1D9B SQSTM1 MIR1229 RNF130 |
esv3607755 | Chr.5:179853436 - 179869950 on Build GRCh38 | Loss | LOC100996419 C5orf45 TBC1D9B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |