Assay Details
Target Gene Details
Entrez Gene ID: | 84250 |
Gene Name: | SMC5-SMC6 complex localization factor 1 |
Gene Aliases: |
ANKRD32, BRCTD1, BRCTx |
Location: |
Chr.5:94613477-94697733 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLF1 | NM_032290.3 | NP_115666.2 | ||
XM_017009979.1 | XP_016865468.1 | |||
XM_017009981.1 | XP_016865470.1 | |||
AK297452.1 | ||||
AK299080.1 | ||||
AL050298.1 | CAB43397.2 | |||
BC054885.1 | AAH54885.2 | |||
BC070332.1 | AAH70332.1 | |||
BC107571.1 | AAI07572.1 | |||
CR936768.1 | ||||
DR001089.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv462254 | Chr.5:94621232 - 94721120 on Build GRCh38 | Loss | SLF1 MCTP1 |
esv2759354 | Chr.5:94429500 - 94706496 on Build GRCh38 | Loss | SLF1 MCTP1 KIAA0825 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |