Assay Details
Target Gene Details
Entrez Gene ID: | 821 |
Gene Name: | calnexin |
Gene Aliases: |
CNX, IP90, P90 |
Location: |
Chr.5:179678631-179731641 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CANX | XM_011534665.2 | XP_011532967.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv969028 | Chr.5:179690223 - 179696616 on Build GRCh38 | Duplication | CANX |
dgv214n111 | Chr.5:179620185 - 179696634 on Build GRCh38 | Duplication | CBY3 HNRNPH1 LOC105377763 CANX C5orf60 |
nsv830560 | Chr.5:179612211 - 179748757 on Build GRCh38 | Loss | CBY3 HNRNPH1 LOC105377763 MAML1 CANX C5orf60 |
nsv5160 | Chr.5:179675417 - 179709494 on Build GRCh38 | Insertion | CBY3 CANX |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |