Assay Details
Target Gene Details
Entrez Gene ID: | 23244 |
Gene Name: | PDS5 cohesin associated factor A |
Gene Aliases: |
PIG54, SCC-112, SCC112 |
Location: |
Chr.4:39822863-39977956 on Build GRCh38 |
Assay Gene Location: | Within Intron 33 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDS5A | NM_001100399.1 | NP_001093869.1 | ||
XM_011513672.2 | XP_011511974.1 | |||
XM_017007928.1 | XP_016863417.1 | |||
AB014548.1 | BAA31623.1 | |||
AB621803.1 | ||||
AF294791.1 | AAM82347.1 | |||
AK021757.1 | ||||
AK023592.1 | ||||
BC009650.1 | ||||
BC114218.1 | AAI14219.1 | |||
BC126225.1 | ||||
CR749425.1 | CAH18263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422397 | Chr.4:39826035 - 40047821 on Build GRCh38 | Duplication | LOC344967 PDS5A |
nsv461338 | Chr.4:39808834 - 39856900 on Build GRCh38 | Loss | PDS5A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |